missing translation for 'onlineSavingsMsg'
Learn More

3-Nitropyridine, TRC

Product Code. 30469871
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469871 2.5 g 2.5g
30464540 500 mg -
30470295 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469871 Supplier Toronto Research Chemicals Supplier No. TRCN5199002.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-26-9
Molecular Formula C5 H4 N2 O2
Molecular Weight (g/mol) 124.1
Synonym Pyridine, 3-nitro-, 3-Nitropyridine, 5-Nitropyridine
IUPAC Name 3-nitropyridine
SMILES [O-][N+](=O)c1cccnc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C5H4N2O2/c8-7(9)5-2-1-3-6-4-5/h1-4H
Formula Weight 124.0273
Chemical Name or Material 3-Nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.