missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic Acid, TRC

Product Code. 30473294
Change view
Click to view available options
Quantity:
10 g
250 g
Unit Size:
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473294 10 g -
30470307 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30473294 Supplier Toronto Research Chemicals Supplier No. TRCN51529510G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-11-2
Molecular Formula C8 H5 N O6
Molecular Weight (g/mol) 211.13
Synonym 1,2-Benzenedicarboxylic acid, 3-nitro- (9CI, ACI), 3-Nitro-1,2-benzenedicarboxylic acid (ACI), Phthalic acid, 3-nitro- (6CI, 7CI, 8CI), 3-Nitrophthalic acid, m-Nitrophthalic acid, NSC 3120, o-Nitrophthalic acid
IUPAC Name 3-nitrophthalic acid
SMILES OC(=O)c1cccc(c1C(=O)O)[N+](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H5NO6/c10-7(11)4-2-1-3-5(9(14)15)6(4)8(12)13/h1-3H,(H,10,11)(H,12,13)
Formula Weight 211.0117
Chemical Name or Material 3-Nitrophthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.