missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic Acid, Mikromol

Product Code. 30459978
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30459978 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459978 Supplier Mikromol Supplier No. MM3641.300025
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Nitrophthalic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Nitrophthalic Acid
CAS 603-11-2
Analyte or Component Names 3-Nitrophthalic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H5 N O6
Certifications/Compliance ISO 9001
Synonym 1,2-Benzenedicarboxylic acid, 3-nitro- (9CI, ACI), 3-Nitro-1,2-benzenedicarboxylic acid (ACI), Phthalic acid, 3-nitro- (6CI, 7CI, 8CI), 3-Nitrophthalic acid, m-Nitrophthalic acid, NSC 3120, o-Nitrophthalic acid
InChI Formula InChI=1S/C8H5NO6/c10-7(11)4-2-1-3-5(9(14)15)6(4)8(12)13/h1-3H,(H,10,11)(H,12,13)
Solution Type Neat
SMILES OC(=O)c1cccc(c1C(=O)O)[N+](=O)[O-]
IUPAC Name 3-nitrophthalic acid
Molecular Weight (g/mol) 211.13
Quantity 25 mg
Formula Weight 211.0117 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.