missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic acid, 95%

Product Code. p-7021800
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
This item is not returnable. View return policy

Product Code. 11492437

Brand: Thermo Scientific Alfa Aesar A14281.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 603-11-2 | C8H5NO6 | 211.129 g/mol

3-Nitrophthalic acid can be used as starting reagent in the synthesis of modified 2-iodoxybenzoic acid derivatives. It may be used in the synthesis of brucinium 2-carboxy-6-nitrophthalate dihydrate (systematic name: 2,3-dimethoxy-10-oxostrychnidinium 2-carboxy-6-nitrophthalate dihydrate), via heating with brucine. Clyatt E

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalic acid can be used as starting reagent in the synthesis of modified 2-iodoxybenzoic acid derivatives. It may be used in the synthesis of brucinium 2-carboxy-6-nitrophthalate dihydrate (systematic name: 2,3-dimethoxy-10-oxostrychnidinium 2-carboxy-6-nitrophthalate dihydrate), via heating with brucine. Clyatt E

Solubility
Solubility in methanol is almost transparent. Soluble in 5% water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-11-2
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007138
InChI Key KFIRODWJCYBBHY-UHFFFAOYSA-N
Synonym 3-nitrophthalic acid, 3-nitrobenzene-1,2-dicarboxylic acid, phthalic acid, 3-nitro, 1,2-benzenedicarboxylic acid, 3-nitro, 3-nitro phthalic acid, 3-nitro-phthalic acid, 3-nitrophthalicacid, nitrophthalic acid, 3-nitrophtalic acid, m-nitrophthalic acid
PubChem CID 69043
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point ∼214°C (decomposition)
Quantity 100 g
Beilstein 2054269
Formula Weight 211.13
Percent Purity 95%
Chemical Name or Material 3-Nitrophthalic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.