missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophthalic acid, 95%

Product Code. 10408340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10408340 500 g Plastic bottle 500g
10235730 100 g Plastic bottle 100g
11945051 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10408340 Supplier Thermo Scientific Acros Supplier No. 160165000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 603-11-2 | C8H5NO6 | 211.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-11-2
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.13
InChI Key KFIRODWJCYBBHY-UHFFFAOYSA-N
Synonym 3-nitrophthalic acid, 3-nitrobenzene-1,2-dicarboxylic acid, phthalic acid, 3-nitro, 1,2-benzenedicarboxylic acid, 3-nitro, 3-nitro phthalic acid, 3-nitro-phthalic acid, 3-nitrophthalicacid, nitrophthalic acid, 3-nitrophtalic acid, m-nitrophthalic acid
PubChem CID 69043
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point 213.0°C to 216.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 09,823
Solubility Information Solubility in water: 20g/L (25 c). Other solubilities: 1g/10mL methanol
Formula Weight 211.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Nitrophthalic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrophthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.