missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylacetic acid, 99%, Thermo Scientific™

Product Code. 10468100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10468100 1 g Glass bottle 1g
10627124 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10468100 Supplier Thermo Scientific Acros Supplier No. 147050010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1877-73-2
MDL Number MFCD00007278
InChI Key WUKHOVCMWXMOOA-UHFFFAOYSA-N
Synonym 3-nitrophenylacetic acid, 2-3-nitrophenyl acetic acid, 3-nitrophenyl acetic acid, benzeneacetic acid, 3-nitro, m-nitrophenyl acetic acid, m-nitrophenylacetic acid, 3-nitrobenzeneacetic acid, acetic acid, m-nitrophenyl, ccris 2337
PubChem CID 15876
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 116.0°C to 120.0°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7NO4
Linear Formula O2NC6H4CH2CO2H
Quantity 1 g
Beilstein 09, 455
Molecular Weight (g/mol) 181.147
Formula Weight 181.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.