missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenyl Ethyl(methyl)carbamate, TRC

Artikelnummer. 30468003
Ansicht ändern
Click to view available options
Quantity:
100 mg
1 g
10 g
Packungsgröße:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
30468003 1 g -
30473511 100 mg -
30464457 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30468003 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCN5034701G
 Restricted Item
Estimated Shipment: 09-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346242-31-6
Molecular Formula C10 H12 N2 O4
Molecular Weight (g/mol) 224.21
Synonym Carbamic acid, N-ethyl-N-methyl-, 3-nitrophenyl ester, 3-Nitrophenyl Ethyl(methyl)carbamate
IUPAC Name (3-nitrophenyl) N-ethyl-N-methylcarbamate
SMILES CCN(C)C(=O)Oc1cccc(c1)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H12N2O4/c1-3-11(2)10(13)16-9-6-4-5-8(7-9)12(14)15/h4-7H,3H2,1-2H3
Formula Weight 224.0797
Chemical Name or Material 3-Nitrophenyl Ethyl(methyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.