missing translation for 'onlineSavingsMsg'
Learn More

(3-Nitrophenyl)acetic Acid, TRC

Product Code. 30475061
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475061 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475061 Supplier Toronto Research Chemicals Supplier No. TRCN5022251G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-Nitrophenyl)acetic Acid
CAS 1877-73-2
Molecular Formula C8 H7 N O4
Quantity 1 g
Synonym Benzeneacetic acid','3-nitro-, Acetic acid','(m-nitrophenyl)- (6CI,7CI,8CI), 3-Nitrobenzeneacetic acid, (m-Nitrophenyl)acetic acid, 2-(3-Nitrophenyl)acetic acid, 3-Nitrophenylacetic acid, NSC 93911
InChI Formula InChI=1S/C8H7NO4/c10-8(11)5-6-2-1-3-7(4-6)9(12)13/h1-4H,5H2,(H,10,11)
SMILES OC(=O)Cc1cccc(c1)[N+](=O)[O-]
IUPAC Name 2-(3-nitrophenyl)acetic acid
Molecular Weight (g/mol) 181.15
Formula Weight 181.0375
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.