missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzylamine hydrochloride, 97%

Product Code. 10347700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347700 1 g Glass bottle 1g
10752611 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347700 Supplier Thermo Scientific Acros Supplier No. 184730010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26177-43-5 | C7H8N2O2·HCl | 188.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26177-43-5
Molecular Formula C7H8N2O2·HCl
Molecular Weight (g/mol) 188.62
MDL Number MFCD00012858
InChI Key DLZXLCHQWOZGSE-UHFFFAOYSA-N
Synonym 3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
PubChem CID 2724030
IUPAC Name (3-nitrophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 226°C to 231°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula O2NC6H4CH2NH2·HCl
Quantity 1 g
Beilstein 12, 1076
Formula Weight 188.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Nitrobenzylamine hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrobenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.