missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl chloride, 99%, Thermo Scientific™

Product Code. 10177460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177460 100 g Glass bottle 100g
10229930 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177460 Supplier Thermo Scientific Acros Supplier No. 404901000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 619-23-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007272
InChI Key APGGSERFJKEWFG-UHFFFAOYSA-N
Synonym 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
PubChem CID 12078
IUPAC Name 1-(chloromethyl)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(CCl)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 41°C to 46°C
Color Brown to Yellow
Boiling Point 85°C to 87°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula O2NC6H4CH2Cl
Quantity 100 g
Formula Weight 171.58
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 3-Nitrobenzyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.