missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl chloride, 98%

Product Code. 11452798
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452798 10 g 10g
11462798 50 g 50g
11472798 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452798 Supplier Thermo Scientific Alfa Aesar Supplier No. A16775.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-23-8 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-23-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007272
InChI Key APGGSERFJKEWFG-UHFFFAOYSA-N
Synonym 3-nitrobenzyl chloride, 1-chloromethyl-3-nitrobenzene, m-nitrobenzyl chloride, alpha-chloro-3-nitrotoluene, benzene, 1-chloromethyl-3-nitro, toluene, alpha-chloro-m-nitro, .alpha.-chloro-m-nitrotoluene, unii-ojl133898o, 3-nitrobenzylchloride, ccris 2324
PubChem CID 12078
IUPAC Name 1-(chloromethyl)-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(CCl)=C1

Specifications

Melting Point 43°C to 47°C
Boiling Point 150°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3261
Beilstein 742794
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.