missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Nitrobenzyl bromide, 98+%

Artikelnummer. 10052670 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
25 g
Packaging:
Glass bottle
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
10052670 5 g Glass bottle 5g
10036093 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10052670 Leverandør Thermo Scientific Acros Leverandørnr. 206920050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 3958-57-4 | C7H6BrNO2 | 216.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 3958-57-4
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007271
InChI Key LNWXALCHPJANMJ-UHFFFAOYSA-N
Synonym 3-nitrobenzyl bromide, 1-bromomethyl-3-nitrobenzene, m-nitrobenzyl bromide, benzene, 1-bromomethyl-3-nitro, alpha-bromo-3-nitrotoluene, m-bromomethyl nitrobenzene, alpha-bromo-m-nitrotoluene, m-nitro-alpha-bromotoluene, 3-nitrobenzylbromide, toluene, alpha-bromo-m-nitro
PubChem CID 77568
IUPAC Name 1-(bromomethyl)-3-nitrobenzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CBr

Tekniske data

CAS Min % 98.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color Yellow to Brown
Boiling Point 153°C to 154°C (7.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2Br
Quantity 5 g
Beilstein 05, 334
Solubility Information Solubility in water: insoluble
Formula Weight 216.03
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Nitrobenzyl bromide
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 3-Nitrobenzyl bromide
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.