missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl alcohol, 99%

Product Code. 11440750
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440750 5 g 5g
11430750 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440750 Supplier Thermo Scientific Alfa Aesar Supplier No. 044445.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-25-0 | C7H7NO3 | 153.137 g/mol

3-Nitrobenzyl alcohol is the best substrate for cytosolic alcohol dehydrogenase. 3-Nitrobenzyl alcohol was employed as matrix during fast-atom bombardment mass spectrometry. It was used in isolation of palmitylated peptide fragment from bovine rhodopsin and its characterization by mass spectrometry. It has been used as a liquid matrix for fast atom bombardment[1] and matrix-assisted laser desorption ionization. In electrospray ionization 3-NBA is doped into low surface tension spray solvents to increase analyte charging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-25-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007273
InChI Key CWNPOQFCIIFQDM-UHFFFAOYSA-N
Synonym 3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
PubChem CID 69267
IUPAC Name (3-nitrophenyl)methanol
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CO

Specifications

Melting Point 29°C to 33°C
Color Yellow
Density 1.300 g/mL
Boiling Point 175°C to 180°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.573
Quantity 5 g
Beilstein 2044769
Solubility Information It is soluble in water.
Formula Weight 153.14
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 3-Nitrobenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.