missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl alcohol, 98%

Product Code. 11417297
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417297 100 g 100g
11407297 25 g 25g
11427297 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417297 Supplier Thermo Scientific Alfa Aesar Supplier No. A15040.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-25-0 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-25-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007273
InChI Key CWNPOQFCIIFQDM-UHFFFAOYSA-N
Synonym 3-nitrobenzyl alcohol, 3-nitrophenyl methanol, m-nitrobenzyl alcohol, benzenemethanol, 3-nitro, benzyl alcohol, m-nitro, 3-nitrobenzenemethanol, unii-f829x990iv, ccris 7971, 3-nitrophenyl methan-1-ol, 3-nitrobenzylalcohol
PubChem CID 69267
IUPAC Name (3-nitrophenyl)methanol
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CO

Specifications

Melting Point 29°C to 33°C
Density 1.3
Boiling Point 175°C to 180°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5755
Quantity 100 g
Beilstein 2044769
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.