missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoyl chloride, 98%

Product Code. 11483234
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483234 50 g 50g
11493234 250 g 250g
11473234 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483234 Supplier Thermo Scientific Alfa Aesar Supplier No. A10579.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-90-4 | C7H4ClNO3 | 185.563 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-90-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.563
MDL Number MFCD00007247
InChI Key NXTNASSYJUXJDV-UHFFFAOYSA-N
Synonym m-nitrobenzoyl chloride, benzoyl chloride, 3-nitro, benzoyl chloride, m-nitro, 3-nitro-benzoyl chloride, ccris 1186, m-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove, 3-nitrobenzoylchloride, chlorid kyseliny m-nitrobenzoove czech, m-nitrobenzoylchlorid
PubChem CID 8495
IUPAC Name 3-nitrobenzoyl chloride
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)Cl

Especificaciones

Melting Point 31°C to 35°C
Density 1.42
Boiling Point 276°C to 278°C (decomposition)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 50 g
UN Number UN3261
Beilstein 777186
Sensitivity Moisture sensitive
Formula Weight 185.57
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzoyl chloride
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • 3-Nitrobenzoyl chloride
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: 3-Nitrobenzoyl chloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.