missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzotrifluoride, 97%

Product Code. 11322306
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322306 100 g 100g
11312306 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322306 Supplier Thermo Scientific Alfa Aesar Supplier No. L05948.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-46-4 | C7H4F3NO2 | 191.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-46-4
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007260
InChI Key WHNAMGUAXHGCHH-UHFFFAOYSA-N
Synonym 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
PubChem CID 7386
IUPAC Name 1-nitro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point -5°C
Density 1.435
Boiling Point 201°C to 205°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.472
Quantity 100 g
UN Number UN2306
Beilstein 2051362
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 3-Nitrobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.