missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzonitrile, 98%

Product Code. 11492488
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
Unit Size:
10g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492488 10 g 10g
11359459 25 g 25g
11402498 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492488 Supplier Thermo Scientific Alfa Aesar Supplier No. A15821.09
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 619-24-9 | C7H4N2O2 | 148.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-24-9
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD00007194
InChI Key RUSAWEHOGCWOPG-UHFFFAOYSA-N
Synonym benzonitrile, 3-nitro, m-nitrobenzonitrile, 3-cyanonitrobenzene, m-cyanonitrobenzene, benzonitrile, m-nitro, 3-cyano-1-nitrobenzene, 3-nitro-benzonitrile, ccris 2327, 3-nitrobenzenecarbonitrile, 3-nitrobenzonitril
PubChem CID 12079
IUPAC Name 3-nitrobenzonitrile
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 114°C to 117°C
Boiling Point 165°C (21 mmHg)
Odor Almond-like
Quantity 10 g
UN Number UN3439
Beilstein 637674
Formula Weight 148.12
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.