missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoic acid, 99%

Product Code. 11495583
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495583 1000 g 1000g
11405593 250 g 250g
11415593 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495583 Supplier Thermo Scientific Alfa Aesar Supplier No. A13494.0B
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-92-6 | C7H5NO4 | 167.12 g/mol

3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-92-6
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007251
InChI Key AFPHTEQTJZKQAQ-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid
PubChem CID 8497
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 140°C to 143°C
Density 1.494
Quantity 1000 g
Beilstein 908644
Merck Index 14,6589
Solubility Information Soluble in water. Insoluble in acetic acid.
Formula Weight 167.12
Percent Purity 99%
Chemical Name or Material 3-Nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.