missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzaldehyde, TRC

Product Code. 30134181
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134181 5 g 5g
30137233 10 g 10g
30009522 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134181 Supplier Toronto Research Chemicals Supplier No. TRCN4921355G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-61-6
Molecular Formula C7 H5 N O3
Molecular Weight (g/mol) 151.12
Synonym Benzaldehyde, 3-nitro-, Benzaldehyde, m-nitro- (8CI), 3-Nitrobenzaldehyde, 3-Nitrobenzenecarboxaldehyde, 3-Nitrophenylcarboxaldehyde, 5-Nitrobenzaldehyde, NSC 5504, m-Nitrobenzaldehyde, m-Nitrobenzaldehyde
IUPAC Name 3-nitrobenzaldehyde
SMILES [O-][N+](=O)c1cccc(C=O)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H5NO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-5H
Formula Weight 151.0269
Chemical Name or Material 3-Nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.