missing translation for 'onlineSavingsMsg'
Learn More

3'-Nitroacetophenone, TRC

Product Code. 30180427
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180427 100 g 100g
30179788 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180427 Supplier Toronto Research Chemicals Supplier No. TRCN491440100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
Synonym (3-Nitrophenyl) Methyl Ketone, 1-(3-Nitrophenyl)-1-ethanone, 1-(3-Nitrophenyl)ethanone, 1-Acetyl-3-nitrobenzene, 3-Acetylnitrobenzene, 3'-Nitroacetophenone, Methyl 3-Nitrophenyl Ketone, NSC 5511, m-Acetylnitrobenzene;m-Nitroacetophenone
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)c1cccc(c1)[N+](=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H7NO3/c1-6(10)7-3-2-4-8(5-7)9(11)12/h2-5H,1H3
Formula Weight 165.0426
Chemical Name or Material 3'-Nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.