missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid, TRC

Product Code. 30011410
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011410 50 mg 50mg
30005343 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011410 Supplier Toronto Research Chemicals Supplier No. TRCN49695050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28328-53-2
Molecular Formula C13 H10 N2 O7 S
Molecular Weight (g/mol) 338.29
Synonym Benzoic acid, 3-nitro-4-phenoxy-5-sulfamoyl- (8CI), 3-Nitro-4-phenoxy-5-sulfamoylbenzoic acid
IUPAC Name 3-nitro-4-phenoxy-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)c1cc(cc(c1Oc2ccccc2)[N+](=O)[O-])C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H10N2O7S/c14-23(20,21)11-7-8(13(16)17)6-10(15(18)19)12(11)22-9-4-2-1-3-5-9/h1-7H,(H,16,17)(H2,14,20,21)
Formula Weight 338.0209
Chemical Name or Material 3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.