missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid, Mikromol™

Product Code. 30051294
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051294 25 mg 25mg
17984096 100 mg 100mg
30459963 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051294 Supplier Mikromol Supplier No. MM1290.010025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid
CAS 28328-53-2
Analyte or Component Names 3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H10 N2 O7 S
Certifications/Compliance ISO 9001
Synonym Bumetanide Imp. A (EP), Bumetanide USP Related Compound B, Bumetanide USP RC B, Benzoic acid, 3-nitro-4-phenoxy-5-sulfamoyl- (8CI), 3-Nitro-4-phenoxy-5-sulfamoylbenzoic acid, Bumetanide Related Compound B, Bumetanide Impurity A
InChI Formula InChI=1S/C13H10N2O7S/c14-23(20,21)11-7-8(13(16)17)6-10(15(18)19)12(11)22-9-4-2-1-3-5-9/h1-7H,(H,16,17)(H2,14,20,21)
SMILES NS(=O)(=O)c1cc(cc(c1Oc2ccccc2)[N+](=O)[O-])C(=O)O
IUPAC Name 3-nitro-4-phenoxy-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 338.29
Quantity 25 mg
Formula Weight 338.0209 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.