missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-2,6-lutidine, 99%, Thermo Scientific™

Product Code. 10262562 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10262562 5 g Glass bottle 5g
10068751 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262562 Supplier Thermo Scientific Acros Supplier No. 348150050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15513-52-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00023503
InChI Key AETHUDGJSSKZKT-UHFFFAOYSA-N
PubChem CID 84958
IUPAC Name 2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 32.0°C to 38.0°C
Color Tan to White
Flash Point 37.78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 20, V,6, 52
Synonym 3-nitro-2,6-lutidine, 3-nitro-2,6-dimethylpyridine, pyridine, 2,6-dimethyl-3-nitro, nitrolutidine, pubchem14242, acmc-20a0s3, 2,6-dimethyl-3-nitro-pyridine, pyridine,2,6-dimethyl-3-nitro, 2-methyl-5-nitro-6-methylpyridine
Formula Weight 152.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Nitro-2, 6-lutidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Nitro-2,6-lutidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.