missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-1,8-naphthalic Anhydride, TRC

Product Code. 30473771
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473771 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473771 Supplier Toronto Research Chemicals Supplier No. TRCN4968451G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3027-38-1
Molecular Formula C12H5NO5
Molecular Weight (g/mol) 243.17
Synonym 5-Nitro-1H,3H-Naphtho[1,8-cd]pyran-1,3-dione, 3-Nitro-naphthalic Anhydride, 3-Nitro-1,8-naphthalenedicarboxylic Anhydride, 3-Nitro-1,8-naphthalic Acid Anhydride, 3-Nitronaphthalic Anhydride, 5-Nitro-1H,3H-benzo[de]isochromene-1,3-dione, NSC 32326, YY 187
SMILES [O-][N+](=O)c1cc2C(=O)OC(=O)c3cccc(c1)c23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H5NO5/c14-11-8-3-1-2-6-4-7(13(16)17)5-9(10(6)8)12(15)18-11/h1-5H
Formula Weight 243.0168
Chemical Name or Material 3-Nitro-1,8-naphthalic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.