missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-1,8-naphthalic anhydride, 98%

Product Code. p-174194 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10399390

Brand: Thermo Scientific Acros 278730050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3027-38-1 | C12H5NO5 | 243.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3027-38-1
Molecular Formula C12H5NO5
Molecular Weight (g/mol) 243.17
MDL Number MFCD00006926
InChI Key FLFLZYYDLIKGJQ-UHFFFAOYSA-N
Synonym 3-nitro-1,8-naphthalic anhydride, 3-nitronaphthalic anhydride, naphthalic anhydride, 3-nitro, 5-nitro-1h,3h-benzo de isochromene-1,3-dione, 5-nitrobenzo de isochromene-1,3-dione, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 5-nitro, 11-nitro-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 7-nitro-3-oxatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene-2,4-dione, 5-17-11-00495 beilstein handbook reference
PubChem CID 18194
IUPAC Name 11-nitro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
SMILES [O-][N+](=O)C1=CC2=C3C(C=CC=C3C(=O)OC2=O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 247°C to 254°C
Color Beige to Yellow or Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 523
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,soluble in dmf
Formula Weight 243.18
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Nitro-1, 8-naphthalic anhydride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitro-1,8-naphthalic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.