missing translation for 'onlineSavingsMsg'
Learn More

3-n-Propyl-2-pyrazolin-5-one, 98%

Product Code. 11490708
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11490708 5 g 5g
11400718 25 g 25g
11410718 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11490708

Brand: Thermo Scientific Alfa Aesar B25214.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29211-70-9 | C6H10N2O | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29211-70-9
Molecular Formula C6H10N2O
Molecular Weight (g/mol) 126.16
MDL Number MFCD00051646
InChI Key GVUNLYBSNQOHBD-UHFFFAOYSA-N
PubChem CID 269244
IUPAC Name 3-propyl-1,4-dihydropyrazol-5-one
SMILES CCCC1=NNC(=O)C1

Specifications

Melting Point 206°C to 210°C
Quantity 5 g
Beilstein 878994
Synonym 3-n-propyl-2-pyrazolin-5-one, 5-propyl-2,4-dihydro-3h-pyrazol-3-one, 3-propyl-2-pyrazolin-5-one, 3-propyl-1h-pyrazol-5 4h-one, 3-propyl-4,5-dihydro-1h-pyrazol-5-one, 5-propyl-2,4-dihydropyrazol-3-one, 5-propyl-1,2-dihydro-pyrazol-3-one, 5-propyl-2,4-dihydro-pyrazol-3-one, 3h-pyrazol-3-one,2,4-dihydro-5-propyl, 3h-pyrazol-3-one, 2,4-dihydro-5-propyl
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material 3-n-Propyl-2-pyrazolin-5-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.