missing translation for 'onlineSavingsMsg'
Learn More

3-(N,N-Dimethylpalmitylammonio)propanesulfonate, 98%

Product Code. 15582333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15582333 5 g 5g
15592333 25 g 25g
15502343 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15582333 Supplier Thermo Scientific Acros Supplier No. 458500050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2281-11-0 | C21H45NO3S | 391.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2281-11-0
Molecular Formula C21H45NO3S
Molecular Weight (g/mol) 391.66
MDL Number MFCD00036911
InChI Key TUBRCQBRKJXJEA-UHFFFAOYSA-N
Synonym 3-n,n-dimethylpalmitylammonio propanesulfonate, zwittergent 3-16, 3-hexadecyldimethylammonio propane-1-sulfonate, palmityl sulfobetaine, n-hexadecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-hexadecyl dimethyl ammonio propane-1-sulfonate, 3-palmityldimethylammonio propanesulfonate, 3-hexadecyldimethylammonio propanesulfonate, 3-n,n-dimethylhexadecylammonio propanesulfonate, n-hexadecy-n,n-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 92176
IUPAC Name 3-[hexadecyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Elemental Analysis 3.5 to 3.7% (N, anhydrous) 63.1 to 65.7% (C, anhydrous)
Loss on Drying 2% max.
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: 100g/L.
Formula Weight 391.65
Percent Purity 98%
Chemical Name or Material 3-(N,N-Dimethylpalmitylammonio)propanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.