missing translation for 'onlineSavingsMsg'
Learn More

3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%

Product Code. 10863571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10863571 25 g Glass bottle 25g
10452930 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10863571 Supplier Thermo Scientific Acros Supplier No. 427740250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14933-09-6 | C19H41NO3S | 363.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14933-09-6
Molecular Formula C19H41NO3S
Molecular Weight (g/mol) 363.61
MDL Number MFCD00036910
InChI Key BHATUINFZWUDIX-UHFFFAOYSA-N
Synonym myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate
PubChem CID 84705
ChEBI CHEBI:10129
IUPAC Name 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246.0°C to 248.0°C
Color Blue to Purple
Elemental Analysis 60.57 to 64.96 % C 10.97 to 11.76 % H 3.72 to 3.99 % N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 363.61
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material 3-(N, N-Dimethylmyristylammonio)propanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • 3-(N,N-Dimethylmyristylammonio)propanesulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.