missing translation for 'onlineSavingsMsg'
Learn More

3’-N,N-Di(desmethyl) Azithromycin, TRC

Product Code. 30008667
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008667 50 mg 50mg
17973215 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008667 Supplier Toronto Research Chemicals Supplier No. TRCD44085050MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612069-27-9
Molecular Formula C36 H68 N2 O12
Molecular Weight (g/mol) 720.93
Synonym 1-Oxa-6-azacyclopentadecan-15-one, 11-[(3-amino-3,4,6-trideoxy-β-D-xylo-hexopyranosyl)oxy]-13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(3-Amino-3,4,6-trideoxy-β-D-xylo-hexopyranosyl)oxy]-13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one, Aminoazithromycin A, N',N'-Di(desmethyl)azithromycin, 3'-N,N-Di(desmethyl) Azithromycin, 3'-(N,N-Didemethyl)azithromycin, Aminoazithromycin
IUPAC Name (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-amino-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@H](N)[C@H]3O)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C36H68N2O12/c1-13-25-36(10,44)29(40)22(6)38(11)17-18(2)15-34(8,43)31(50-33-27(39)24(37)14-19(3)46-33)20(4)28(21(5)32(42)48-25)49-26-16-35(9,45-12)30(41)23(7)47-26/h18-31,33,39-41,43-44H,13-17,37H2,1-12H3/t18-,19-,20+,21-,22-,23+,24+,25-,26+,27-,28+,29-,30+,31-,33+,34-,35-,36-/m1/s1
Formula Weight 720.4772
Percent Purity >95%
Chemical Name or Material 3'-N,N-Di(desmethyl) Azithromycin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.