missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(N-Morpholino)ethane Sulfonic Acid (MES), Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
3-(N-Morpholino)ethane Sulfonic Acid (MES)
Specifications
Specifications
| Chemical Name or Material | 3-(N-Morpholino)ethane Sulfonic Acid (MES) |
| CAS | 4432-31-9 |
| Analyte or Component Names | 3-(N-Morpholino)ethane Sulfonic Acid (MES) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H13 N O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-(N-Morpholino)ethane Sulfonic Acid, MES |
| InChI Formula | InChI=1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10) |
| SMILES | OS(=O)(=O)CCN1CCOCC1 |
| IUPAC Name | 2-morpholin-4-ylethanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction