missing translation for 'onlineSavingsMsg'
Learn More

3-[N-Methyl(pentyl)amino]propanenitrile, Mikromol™

Product Code. 30051647
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30051647 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30051647

Brand: Mikromol MM1326.030100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[N-Methyl(pentyl)amino]propanenitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[N-Methyl(pentyl)amino]propanenitrile
CAS 919491-64-8
Analyte or Component Names 3-[N-Methyl(pentyl)amino]propanenitrile
Shipping Condition Room Temperature
Molecular Formula C9 H18 N2
Certifications/Compliance ISO 9001
Synonym 3-(Methylpentylamino)propanenitrile, Propanenitrile, 3-(methylpentylamino)-
InChI Formula InChI=1S/C9H18N2/c1-3-4-5-8-11(2)9-6-7-10/h3-6,8-9H2,1-2H3
SMILES CCCCCN(C)CCC#N
IUPAC Name 3-[methyl(pentyl)amino]propanenitrile
Molecular Weight (g/mol) 154.2526
Quantity 100 mg
Formula Weight 154.147 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.