missing translation for 'onlineSavingsMsg'
Learn More

3-(N-Methyl-N-pentylamino)propionic Acid Hydrochloride, TRC

Product Code. 30136576
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136576 5 g 5g
30136613 10 g 10g
30006173 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136576 Supplier Toronto Research Chemicals Supplier No. TRCM3258765G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625120-81-2
Molecular Formula C9 H19 N O2 . Cl H
Molecular Weight (g/mol) 209.71
Synonym β-Alanine, N-methyl-N-pentyl-, hydrochloride (1:1), β-Alanine, N-methyl-N-pentyl-, hydrochloride (9CI), Ibanic acid hydrochloride, N-Methyl-N-pentyl-β-alanine hydrochloride, 3-(N-Methyl-N-pentylamino)propionic Acid hydrochloride, Ibandronate Sodium Monohydrate Imp. A (EP)
IUPAC Name 3-[methyl(pentyl)amino]propanoic acid;hydrochloride
SMILES Cl.CCCCCN(C)CCC(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H19NO2.ClH/c1-3-4-5-7-10(2)8-6-9(11)12;/h3-8H2,1-2H3,(H,11,12);1H
Formula Weight 209.1183
Percent Purity >95%
Chemical Name or Material 3-[Methyl(pentyl)amino]propanoic Acid Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.