missing translation for 'onlineSavingsMsg'
Learn More

3-N-(Methanesulfonamido)phenylboronic acid, 95%, Thermo Scientific™

Product Code. 15276048 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15276048 1 g 1g
15286048 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15276048 Supplier Thermo Scientific Acros Supplier No. 454250010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 148355-75-3
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.04
InChI Key XUIQQIRLFMCWLN-UHFFFAOYSA-N
Synonym 3-methylsulfonylamino phenylboronic acid, 3-methylsulfonamido phenylboronic acid, 3-methanesulfonylaminophenylboronic acid, 3-methylsulfonylaminophenylboronic acid, 3-methanesulfonamidophenyl boronic acid, 3-methanesulfonamido phenyl boronic acid, 3-methanesulfonamidophenylboronic acid, 3-methylsulphonylamino benzeneboronic acid, n-3-methanesulfonamidephenylboronic acid, 3-methylsulfonylamino benzeneboronic acid
PubChem CID 2773535
IUPAC Name [3-(methanesulfonamido)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)NS(=O)(=O)C)(O)O

Especificaciones

Melting Point 255.0°C to 258.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.04
Percent Purity 95%
Chemical Name or Material 3-N-(Methanesulfonamido)phenylboronic acid
compliance-icons
Nombre del producto
  • 3-N-(Methanesulfonamido)phenylboronic acid, 95%
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.