missing translation for 'onlineSavingsMsg'
Learn More

3' '-[N-(HABA-CBz)] 3,5,6'-Tri(N-Benzyloxycarbonyl) Kanamycin A, TRC

Product Code. 30392807
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392807 5 mg 5mg
30407263 25 mg 25mg
30398696 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392807 Supplier Toronto Research Chemicals Supplier No. TRCH0117555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3''-[N-(HABA-CBz)] 3,5,6'-Tri(N-Benzyloxycarbonyl) Kanamycin A
Analyte or Component Names 3''-[N-(HABA-CBz)] 3,5,6'-Tri(N-Benzyloxycarbonyl) Kanamycin A
Shipping Condition Room Temperature
Molecular Formula C54H67N5O21
InChI Formula InChI=1S/C54H67N5O21/c60-25-38-40(62)39(59-48(68)36(61)21-22-55-51(69)73-26-30-13-5-1-6-14-30)42(64)49(78-38)79-46-34(57-53(71)75-28-32-17-9-3-10-18-32)23-35(58-54(72)76-29-33-19-11-4-12-20-33)47(45(46)67)80-50-44(66)43(65)41(63)37(77-50)24-56-52(70)74-27-31-15-7-2-8-16-31/h1-20,34-47,49-50,60-67H,21-29H2,(H,55,69)(H,56,70)(H,57,71)(H,58,72)(H,59,68)/t34-,35+,36-,37-,38-,39+,40-,41-,42-,43+,44-,45-,46+,47-,49-,50-/m1/s1
Solution Type Neat
SMILES O[C@H]1[C@H](O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CNC(OCC3=CC=CC=C3)=O)O2)[C@@H](NC(OCC4=CC=CC=C4)=O)C[C@@H](NC(OCC5=CC=CC=C5)=O)[C@@H]1O[C@H]6O[C@H](CO)[C@@H](O)[C@H](NC([C@H](O)CCNC(OCC7=CC=CC=C7)=O)=O)[C@H]6O
IUPAC Name dibenzyl ((1S,3R,4S,5R,6R)-4-(((2S,3R,4S,5S,6R)-4-((R)-4-(((benzyloxy)carbonyl)amino)-2-hydroxybutanamido)-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-(((2R,3R,4S,5S,6R)-6-((((benzyloxy)carbonyl)amino)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-5-hydroxycyclohexane-1,3-diyl)dicarbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 1122.13
Quantity 5 mg
Formula Weight 1141.43
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.