missing translation for 'onlineSavingsMsg'
Learn More

3-n-Decylthiophene, 97%, Thermo Scientific™

Product Code. 15491428
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15491428 5 g 5g
15481428 1 g 1g
15401438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491428 Supplier Thermo Scientific Alfa Aesar Supplier No. H55251.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65016-55-9
Molecular Formula C14H24S
Molecular Weight (g/mol) 224.406
MDL Number MFCD00143180
InChI Key JAYBIBLZTQMCAY-UHFFFAOYSA-N
Synonym 3-n-decylthiophene, 3-decylthiophene, thiophene, 3-decyl, acmc-1bell, ksc491i4d
PubChem CID 566842
IUPAC Name 3-decylthiophene
SMILES CCCCCCCCCCC1=CSC=C1

Specifications

Density 0.912
Boiling Point 119°C to 120°C (1.2 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.489
Quantity 5 g
Formula Weight 224.4
Percent Purity 97%
Chemical Name or Material 3-n-Decylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.