missing translation for 'onlineSavingsMsg'
Learn More

3’N-Benzyl Biotin, TRC

Product Code. 30011360
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011360 50 mg 50mg
30005226 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011360 Supplier Toronto Research Chemicals Supplier No. TRCB23303050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57229-92-2
Molecular Formula C17 H22 N2 O3 S
Molecular Weight (g/mol) 334.43
Synonym (3aS,4S,6aR)-Hexahydro-2-oxo-3-(phenylmethyl)-1H-thieno[3,4-d]imidazole-4-pentanoic Acid, 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-3-(phenylmethyl)-, [3aS-(3aα,4β,6aα)]-, 5-[(3aS,4S,6aR)-1-Benzyl-2-oxohexahydrothieno[3,4-d]imidazol-4-yl]pentanoic Acid
IUPAC Name 5-[(3aS,4S,6aR)-3-benzyl-2-oxo-3a,4,6,6a-tetrahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N(Cc3ccccc3)[C@H]12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H22N2O3S/c20-15(21)9-5-4-8-14-16-13(11-23-14)18-17(22)19(16)10-12-6-2-1-3-7-12/h1-3,6-7,13-14,16H,4-5,8-11H2,(H,18,22)(H,20,21)/t13-,14-,16-/m0/s1
Formula Weight 334.1351
Chemical Name or Material 3'N-Benzyl Biotin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.