missing translation for 'onlineSavingsMsg'
Learn More

[3-(Morpholinomethyl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10187812
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10187812 1 g Amber Glass Bottle 1g
10239252 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10187812 Supplier Thermo Scientific Maybridge Supplier No. CC40109DA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 91271-64-6
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD07772831
InChI Key KFSCOCJCNMSXOK-UHFFFAOYSA-N
Synonym 3-morpholinomethyl phenyl methanol, 3-morpholin-4-ylmethyl phenyl methanol, 3-4-morpholinylmethyl benzenemethanol, 3-morpholin-4-ylmethyl phenyl methan-1-ol
PubChem CID 7162057
IUPAC Name [3-(morpholin-4-ylmethyl)phenyl]methanol
SMILES C1COCCN1CC2=CC(=CC=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material [3-(Morpholinomethyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.