missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholin-4-ylmethyl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10411124
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10411124 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10411124 Supplier Thermo Scientific Maybridge Supplier No. CC40116DA
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 857283-91-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H14N2O
MDL Number MFCD08060511
Quantity 1 g
Synonym 3-morpholin-4-ylmethyl benzonitrile, benzonitrile,3-4-morpholinylmethyl, 3-morpholin-4-yl methyl benzonitrile, 3-morpholinomethyl benzonitrile, 3-morpholin-4-ylmethyl benzenecarbonitrile
InChI Key GSVNKQLSALKJHW-UHFFFAOYSA-N
SMILES C1COCCN1CC2=CC=CC(=C2)C#N
IUPAC Name 3-(morpholin-4-ylmethyl)benzonitrile
Molecular Weight (g/mol) 202.257
PubChem CID 7164605
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)benzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.