missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate, 97%, Thermo Scientific™

Product Code. 10729983
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10729983 1 g 1g
10527663 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10729983 Supplier Thermo Scientific Maybridge Supplier No. CC40151DA
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 137605-80-2
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.71
MDL Number MFCD09064949
InChI Key UOMMCPQFAXOMMO-UHFFFAOYSA-N
Synonym 3-morpholin-4-ylmethyl benzoic acid hydrochloride, 3-morpholinomethyl benzoic acid hydrochloride, 3-morpholin-4-ylmethyl benzoic acid, chloride, 3-morpholin-4-ylmethyl benzoic acid,hydrochloride, 3-morpholin-4-yl methyl benzoic acid hydrochloride, 3-morpholin-4-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 24229486
IUPAC Name 3-(morpholin-4-ylmethyl)benzoic acid;hydrochloride
SMILES Cl.OC(=O)C1=CC(CN2CCOCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 275.74
Percent Purity 97%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.