missing translation for 'onlineSavingsMsg'
Learn More

3-morpholin-4-ylaniline, 97%, Thermo Scientific™

Product Code. 10084612
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10084612 1 g Amber Glass Bottle 1g
10779944 250 mg Amber Glass Bottle 250mg
10173714 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10084612 Supplier Thermo Scientific Maybridge Supplier No. CC33814DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159724-40-0
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD03197165
InChI Key ZJWLMZURLIHVHE-UHFFFAOYSA-N
Synonym 3-4-morpholinyl aniline, 3-morpholinoaniline, 3-morpholinobenzenamine, 3-morpholin-4-yl aniline, benzenamine, 3-4-morpholinyl, 3-morpholin-4-yl-phenylamine, 4-3-aminophenyl morpholine, 3-morpholin-4-ylphenylamine, 3-morpholinylaniline, meta-morpholino aniline
PubChem CID 847768
IUPAC Name 3-morpholin-4-ylaniline
SMILES NC1=CC=CC(=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 3-Morpholin-4-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.