missing translation for 'onlineSavingsMsg'
Learn More

3-Methylvaleric acid, 99%

Product Code. 11493128
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493128 100 g 100g
11473128 5 g 5g
11483128 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493128 Supplier Thermo Scientific Alfa Aesar Supplier No. A18886.22
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 105-43-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-43-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002729,MFCD02262177
InChI Key IGIDLTISMCAULB-UHFFFAOYNA-N
Synonym 3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid
PubChem CID 7755
IUPAC Name 3-methylpentanoic acid
SMILES CCC(C)CC(O)=O

Specifications

Melting Point -42°C
Density 0.93
Boiling Point 196°C to 198°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.416
Quantity 100 g
UN Number UN3265
Beilstein 1720696
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 3-Methylvaleric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.