missing translation for 'onlineSavingsMsg'
Learn More

3-Methyluridine, TRC

Product Code. 30133735
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133735 2.5 mg 2.5mg
30138560 10 mg 10mg
30008402 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133735 Supplier Toronto Research Chemicals Supplier No. TRCM3383552.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2140-69-4
Molecular Formula C10H14N2O6
Molecular Weight (g/mol) 258.23
Synonym N3-Methyluridine, NSC 246077, 3-Methyl-uridine
SMILES O[C@H]1[C@@H](O)[C@H](N2C(N(C)C(C=C2)=O)=O)O[C@@H]1CO

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C10H14N2O6/c1-11-6(14)2-3-12(10(11)17)9-8(16)7(15)5(4-13)18-9/h2-3,5,7-9,13,15-16H,4H2,1H3/t5-,7-,8-,9-/m1/s1
Formula Weight 258.0852
Chemical Name or Material 3-Methyluridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.