missing translation for 'onlineSavingsMsg'
Learn More

Eltoprazine hydrochloride, MedChemExpress

Product Code. 16477128
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16477128

Brand: Medchem Express HY16687A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eltoprazine hydrochloride (DU 28853 hydrochloride) is a serenic or antiaggressive agent which as an agonist at the 5-HT1A and 5-HT1B receptors and as an antagonist at the 5-HT2C receptor.

Eltoprazine hydrochloride (DU 28853 hydrochloride) is a serenic or antiaggressive agent which as an agonist at the 5-HT1A and 5-HT1B receptors and as an antagonist at the 5-HT2C receptor.

Specifications

Chemical Name or Material Eltoprazine hydrochloride
CAS 98206-09-8
Color White
Purity Grade Notes Research
Molecular Formula C12H17ClN2O2
Quantity 10 mg
Synonym DU 28853 hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (389.51 mM) ∣DMSO : ≥ 31 mg/mL (120.75 mM)
SMILES [H]Cl.N1(C2=C3OCCOC3=CC=C2)CCNCC1
Molecular Weight (g/mol) 256.73
Formula Weight 256.73
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.