missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, 99+%

Product Code. 10386860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10386860 50 mL Glass bottle 50mL
10465485 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386860 Supplier Thermo Scientific Acros Supplier No. 127860500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 616-44-4 | C5H6S | 98.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.17
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -69.0°C
Color Colorless to Yellow
Density 1.0160g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5180 to 1.5200
Quantity 50 mL
Beilstein 17, 38
Specific Gravity 1.016
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,aceton and benzene
Formula Weight 98.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.