missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, 98+%

Product Code. 11415403
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415403 100 g 100g
11405403 25 g 25g
11425403 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415403 Supplier Thermo Scientific Alfa Aesar Supplier No. A13394.22
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-44-4 | C5H6S | 98.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005470
InChI Key QENGPZGAWFQWCZ-UHFFFAOYSA-N
Synonym thiophene, 3-methyl, 3-thiotolene, beta-methylthiophene, poly, p3mt, 3-methyl thiophene, 3-methyl-thiophene, methyl-3-thiophene, ccris 2937, thiophene, 3-methyl-, homopolymer
PubChem CID 12024
IUPAC Name 3-methylthiophene
SMILES CC1=CSC=C1

Specifications

Melting Point -69°C
Density 1.02
Boiling Point 115°C to 117°C
Flash Point 11°C (51°F)
Odor Aromatic
Refractive Index 1.52
Quantity 100 g
UN Number UN1993
Beilstein 1300
Formula Weight 98.17
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methylthiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Methylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.