missing translation for 'onlineSavingsMsg'
Learn More

3-methylthiophene-2-carbonyl Chloride, 98%, Thermo Scientific™

Product Code. 10197832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197832 2 g Glass bottle 2g
10556933 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197832 Supplier Thermo Scientific Acros Supplier No. 380400020
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 61341-26-2
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00130091
InChI Key AJSVPEVDFBYRCH-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
PubChem CID 2737605
IUPAC Name 3-methylthiophene-2-carbonyl chloride
SMILES CC1=C(SC=C1)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 216°C to 220°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5830 to 1.585
Quantity 2 g
Formula Weight 160.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene-2-carbonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylthiophene-2-carbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.