missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylthio)propylamine, TRC
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Specifications
Specifications
| Chemical Name or Material | 3-(Methylthio)propylamine |
| CAS | 4104-45-4 |
| Molecular Formula | C4H11NS |
| Quantity | 500 mg |
| Synonym | 1-Propanamine','3-(methylthio)- (9CI','ACI), 3-(Methylthio)-1-propanamine (ACI), Propylamine','3-(methylthio)- (6CI','7CI','8CI), 1-Amino-3-(methylthio)propane, 3-(Methylmercapto)propylamine, 3-(Methylsulfanyl)propylamine, 3-(Methylthio)-1-propylamine, 3-(Methylthio)propylamine, 3-Aminopropyl methyl sulfide, 3-Methylsulfanyl-1-propylamine, 3-Methylsulfanylpropan-1-amine, S-Methylhomocysteamine |
| InChI Formula | InChI=1S/C4H11NS/c1-6-4-2-3-5/h2-5H2,1H3 |
| SMILES | CSCCCN |
| IUPAC Name | 3-methylsulfanylpropan-1-amine |
| Molecular Weight (g/mol) | 105.2 |
| Formula Weight | 105.0612 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction