missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propylamine, 97%, Thermo Scientific™

Product Code. 10493433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10493433 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10493433 Supplier Thermo Scientific Acros Supplier No. 414930010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4104-45-4
CAS Min % 96.0
CAS Max % 100.0
Density 0.9600g/mL
Boiling Point 169.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H11NS
Linear Formula CH3SCH2CH2CH2NH2
Refractive Index 1.4920 to 1.4940
MDL Number MFCD00041898
Quantity 1g
Beilstein 04, IV, 1655
Specific Gravity 0.96
Synonym 3-methylthiopropylamine, 3-methylthio propylamine, 1-propanamine, 3-methylthio, 3-methylthiopropanamine, methylmercaptopropylamine, unii-1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylthio-1-propanamine, 3-methylsulfanyl propan-1-amine, 1iap0cgd5s
InChI Key KKYSBGWCYXYOHA-UHFFFAOYSA-N
SMILES CSCCCN
IUPAC Name 3-methylsulfanylpropan-1-amine
Molecular Weight (g/mol) 105.2
PubChem CID 77743
ChEBI CHEBI:16696
Formula Weight 105.2
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)propylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Methylthio)propylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.