missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11419677
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419677 10 g 10g
11429677 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419677 Supplier Thermo Scientific Alfa Aesar Supplier No. B22150.09
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 505-79-3 | Molecular Formula: C5H9NS2 | Molecular Weight: 147.254 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-79-3
Molecular Formula C5H9NS2
Molecular Weight (g/mol) 147.254
MDL Number MFCD00209521
InChI Key LDKSCZJUIURGMW-UHFFFAOYSA-N
Synonym 3-methylthio propyl isothiocyanate, iberverin, 3-methylthiopropyl isothiocyanate, ibervirin, propane, 1-isothiocyanato-3-methylthio, unii-ytt13bn94z, 3-methylmercaptopropyl isothiocyanate, isothiocyanic acid, 3-methylthio propyl ester, 1-isothiocyanato-3-methylthio propane, fema no. 3312
PubChem CID 62351
IUPAC Name 1-isothiocyanato-3-methylsulfanylpropane
SMILES CSCCCN=C=S

Specifications

Density 1.113
Boiling Point 254°C
Flash Point 110°C (230°F)
Odor Pungent
Refractive Index 1.564
Quantity 10 g
UN Number UN2922
Sensitivity Moisture sensitive
Formula Weight 147.25
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.