missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)propionic acid, MedChemExpress

Product Code. 16404596
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16404596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16404596 Supplier Medchem Express Supplier No. HY101401
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 3-(Methylthio)propionic acid is an intermediate in the methionine metabolism.

3-(Methylthio)propionic acid is an intermediate in the methionine metabolism.

Specifications

Chemical Name or Material 3-(Methylthio)propionic acid
CAS 646-01-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C4H8O2S
Synonym 3-Methylsulfanylpropionic acid
Solubility Information H2O : 25 mg/mL (208.04 mM; Need ultrasonic)
SMILES O=C(O)CCSC
Molecular Weight (g/mol) 120.17
Quantity 100 mg
Formula Weight 120.17
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.